missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11398727

Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11398727
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11398727

Brand: Thermo Scientific Alfa Aesar 035834.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bismuth(III) sulfide is used as a starting material to produce other bismuth compounds. It is used in glass industry as a colorant, electronic ceramics, scintillator BGO and in superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-07-9
Molecular Formula Bi2S3
Molecular Weight (g/mol) 514.14
MDL Number MFCD00014204
InChI Key YNRGZHRFBQOYPP-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Bi+3].[Bi+3]

Specifications

Color Black-Brown
Physical Form Crystals and/or Chunks
Quantity 50 g
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 514.15
Percent Purity 99.9%
Density 7.7 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ24K056 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number035834.30,035834.18
Lot NumberR06K023 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog Number035834.30,035834.18
Lot NumberS12J029 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog Number035834.30,035834,035834.18
Lot NumberM23J015 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number035834.30,035834.18
Lot NumberU10I001 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog Number035834.30,035834.18
compliance-icons
Product Identifier
  • Bismuth(III) sulfide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.