Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
£184.00 - £690.00
Specifications
Material Phenolic Resin
Closure Color Black
Closure Liner Polyseal
Closure Material Phenolic
Closure Type Screw Cap
View More Specs
Products 8
Product Code Brand dimClosureSize Quantity Price Quantity & Availability  
Product Code Brand dimClosureSize Quantity Price Quantity & Availability  
11538172
View Documents
Fisherbrand™
02911803
5500/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15592351
View Documents
Fisherbrand™
02912055
576/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15763575
View Documents
Fisherbrand™
02911806A
4100/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11598172
View Documents
Fisherbrand™
02912101
576/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15512361
View Documents
Fisherbrand™
02912062
576/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15715998
View Documents
Fisherbrand™
02912091A
2200/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11538182
View Documents
Fisherbrand™
02911727
1600/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15573261
View Documents
Fisherbrand™
02912063
576/Cs. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Specifications
Specifications
Phenolic Resin
Polyseal
Screw Cap
Black
Phenolic