Learn More
Blasticidin S Hydrochloride (From Streptomyces Griseochromogenes), Fisher BioReagents
£235.00 - £526.00
Chemical Identifiers
CAS | 3513-03-9 |
---|---|
Molecular Formula | C17H27ClN8O5 |
Molecular Weight (g/mol) | 458.90 |
InChI Key | YQXYQOXRCNEATG-UHFFFAOYNA-N |
Synonym | BLA-S, Cytovirin |
PubChem CID | 124080960 |
IUPAC Name | (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride |
SMILES | CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10264913
|
Fisher BioReagents
BP2647-25 |
25 mg | Amber Glass |
£235.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10658203
|
Fisher BioReagents
BP2647-50 |
50 mg | Amber Glass |
£270.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10648203
|
Fisher BioReagents
BP2647-100 |
100 mg | Amber Glass |
£526.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
3513-03-9 | |
458.90 | |
BLA-S, Cytovirin | |
(2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride |
C17H27ClN8O5 | |
YQXYQOXRCNEATG-UHFFFAOYNA-N | |
124080960 | |
CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl |
Specifications
Blasticidin S Hydrochloride | |
3513-03-9 | |
≥99.0 % | |
900μg/mg min. | |
YQXYQOXRCNEATG-UHFFFAOYNA-N | |
(2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride | |
124080960 | |
99.0% min. | |
Blasticidin S Hydrochloride, From Streptomyces griseochromogenes |
Powder/Solid | |
248°C | |
C17H27ClN8O5 | |
BLA-S, Cytovirin | |
CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl | |
458.90 | |
458.69 | |
White |
Safety and Handling
DANGER!
Emergency Overview
May be fatal if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:4
Flammability:0
Instability:0
Recommended Storage : 0° to 5°C
- Blasticidin S Hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.