missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 193885, Tocris Bioscience™
Potent, competitive NPY Y1 receptor antagonist
£252.32 - £1015.92
Chemical Identifiers
CAS | 679839-66-8 |
---|---|
Molecular Formula | C36H48N4O9 |
Molecular Weight (g/mol) | 680.80 |
MDL Number | MFCD16878998 |
InChI Key | BYHJIPSVXAFCDI-UHFFFAOYNA-N |
Synonym | 1,4-dihydro-4-3-3-4-3-methoxyphenyl-1-piperidinyl propyl amino carbonyl amino phenyl-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester l-lactate |
PubChem CID | 56972234 |
IUPAC Name | 2-hydroxypropanoic acid; 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
SMILES | CC(O)C(O)=O.COC(=O)C1=C(C)NC(C)=C(C1C1=CC(NC(=O)NCCCN2CCC(CC2)C2=CC(OC)=CC=C2)=CC=C1)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18054652
|
Tocris Bioscience
3242/10 |
10 mg |
£252.32
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18085602
|
Tocris Bioscience
3242/50 |
50 mg |
£1015.92
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, competitive neuropeptide (NPY) Y1 antagonist (Ki = 3.3 nM, IC50 = 5.9 nM) that displays > 47, > 100, > 160, > 160 and > 160-fold selectivity over σ1, α1, Y2, Y4 and Y5 receptors respectively. Reduces food intake and body weight via central Y1 inhibition and is brain penetrant.Chemical Identifiers
679839-66-8 | |
680.80 | |
BYHJIPSVXAFCDI-UHFFFAOYNA-N | |
56972234 | |
CC(O)C(O)=O.COC(=O)C1=C(C)NC(C)=C(C1C1=CC(NC(=O)NCCCN2CCC(CC2)C2=CC(OC)=CC=C2)=CC=C1)C(=O)OC |
C36H48N4O9 | |
MFCD16878998 | |
1,4-dihydro-4-3-3-4-3-methoxyphenyl-1-piperidinyl propyl amino carbonyl amino phenyl-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester l-lactate | |
2-hydroxypropanoic acid; 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
Specifications
679839-66-8 | |
MFCD16878998 | |
BYHJIPSVXAFCDI-UHFFFAOYNA-N | |
2-hydroxypropanoic acid; 3,5-dimethyl 4-{3-[({3-[4-(3-methoxyphenyl)piperidin-1-yl]propyl}carbamoyl)amino]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate | |
56972234 | |
>99% |
C36H48N4O9 | |
1,4-dihydro-4-3-3-4-3-methoxyphenyl-1-piperidinyl propyl amino carbonyl amino phenyl-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester l-lactate | |
CC(O)C(O)=O.COC(=O)C1=C(C)NC(C)=C(C1C1=CC(NC(=O)NCCCN2CCC(CC2)C2=CC(OC)=CC=C2)=CC=C1)C(=O)OC | |
680.80 | |
680.79 | |
BMS 193885 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature