Learn More
Lovibond™ BOD Sample bottle
For OxiDirect and BD 600
Brand:  Lovibond™ 418645
Product Code. 10121682
  • £51.15 / Pack of 6
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For OxiDirect and BD 600
Sample bottle 500mL for BOD apparatus 6pcs brown glass For OxiDirect and BD 600. 
Specifications
Specifications
Sample Bottle
Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ BOD Sample bottle >