Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Keeps test tubes upright and secure in the bath.
Brand:  Grant Instruments™ J2-25
 View more versions of this product
Product Code. 11446397
  • £124.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Keeps test tubes upright and secure in the bath.
  • Compatible with a wide range of Grant Unstirred Water Baths
  • Holds up to 105 test tubes depending on size
  • Dual slotted trays keep tubes secure and prevent tipping
TRUSTED_SUSTAINABILITY
Specifications
Specifications
18 25 mm Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Test Tube Rack for Unstirred Water Baths > Capacity: 18 Tubes