missing translation for 'onlineSavingsMsg'
Learn More
Boron nitride paste, Thermo Scientific Chemicals Product Code.: 11350408

Boron nitride paste, Thermo Scientific Chemicals

Product Code. 11350408
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11350408

Brand: Thermo Scientific Alfa Aesar 043773.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength

Resistance against molten metals and salts, high thermal conductivity and dielectric strength

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Color White
Physical Form Paste
Quantity 25 g
IUPAC Name azanylidyneborane
Odor Odorless
Appearance White paste
Density 1.4
Chemical Name or Material Boron nitride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27L039 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number043773.22,043773.14,043773.36,043773.A3
Lot NumberN03L003 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number043773.22,043773.14,043773.36,043773.A3
Lot NumberS02J023 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number043773.22,043773.14,043773.36
Lot NumberY04I029 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number043773.22,043773.14,043773.36
Lot NumberZ21H025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043773.22,043773.14,043773.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron nitride paste, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.