missing translation for 'onlineSavingsMsg'
Learn More
Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11358938

Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11358938
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11358938

Brand: Thermo Scientific Alfa Aesar 041786.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500gShow MoreShow Less
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo[(oxoboranyl)oxy]borane
SMILES O=BOB=O

Specifications

Boiling Point 1860°C
Melting Point ∽450°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.999%
Solubility Information Soluble in water. Slightly soluble in methanol.
Formula Weight 69.62
Odor Odorless
Sensitivity Hygroscopic
Density 2.46
Chemical Name or Material Boron oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM30K025 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number041786.18,041786.09,041786.04
Lot NumberS21I017 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog Number041786.18,041786.09,041786.04
Lot NumberY19H014 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041786.18,041786.09,041786.04
Lot NumberP09I005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041786.18,041786.09,041786.04
Lot NumberX22G013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041786.18,041786.09,041786.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.