missing translation for 'onlineSavingsMsg'
Learn More
Boron trifluoride dihydrate, 65% BF3, Thermo Scientific Chemicals Product Code.: 10539423

Boron trifluoride dihydrate, 65% BF3, Thermo Scientific Chemicals

Product Code. 10539423
500 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10539423

Brand: Thermo Scientific Acros 388715000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13319-75-0
Molecular Formula BF3H4O2
Molecular Weight (g/mol) 103.84
MDL Number MFCD00149901
InChI Key MJCYPBSRKLJZTB-UHFFFAOYSA-N
Synonym boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058Show MoreShow Less
PubChem CID 61591
IUPAC Name trifluoroborane dihydrate
SMILES O.O.FB(F)F

Specifications

Boiling Point 58°C to 60°C (1.5mmHg)
Melting Point 6°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 65.0 to 66%
Linear Formula BF3·2H2O
Solubility Information Solubility in water: miscible
Formula Weight 103.85
Percent Purity 65% (BF3)
Identification Confirmed
Packaging Glass bottle
Specific Gravity 1.642
Density 1.6420g/mL
Chemical Name or Material Boron trifluoride dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0422150 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number388711000,388715000
Lot NumberA0448220 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number388711000,388715000
Lot NumberA0325839 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number388711000,388715000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron trifluoride dihydrate, 65% BF3, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.