Learn More
Description
Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 210°C |
Melting Point | 48°C to 51°C |
Color | White |
Physical Form | Crystals |
Quantity | 5 g |
Assay Percent Range | >98% |
Solubility Information | Reacts with water. |
Formula Weight | 391.52 |
Odor | Pungent |
Sensitivity | Light and moisture sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X28K010 | Certificate of Analysis | 16/12/2024 | 087220.06,087220.03,087220.14 |
S28K019 | Certificate of Analysis | 25/07/2024 | 087220.06,087220.03,087220.14 |
Y16J024 | Certificate of Analysis | 15/12/2023 | 087220.06,087220.03,087220.14 |
R12J036 | Certificate of Analysis | 25/05/2023 | 087220.06,087220.03 |
P16I052 | Certificate of Analysis | 12/04/2022 | 087220.06,087220.03 |
Safety and Handling


- Boron triiodide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Boron triiodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.