Learn More
Soda Lime Glass Petri Dishes
High-quality petri dish, with lid.
Brand: Fisherbrand 2290/80X15
Description
- Constructed of soda lime glass.
- Includes lid
Specifications
Closure Included | |
Round | |
15 mm |
Soda Lime Glass | |
Petri Dish | |
80 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.