missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispensette™ Bottletop Dispenser Discharge Tube

For use with the Dispensette III™ and Organic dispensers, for dispensing aggressive and organic solvents, respectively.

Brand:  BRAND™ 707918

 View more versions of this product

Product Code. 10675581

  • £47.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With integrated valve
  • Provided individually
  • Fine tip shape

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fine tip
25, 50, 100 mL
1/Pk.
Replacement Discharge Tube
25, 50, 100mL Fine tip Dispensette™ III bottletop dispenser
120 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Dispensette™ Bottletop Dispenser Discharge Tube > Fine Tip, 25, 50, 100mL; for Dispensette III™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.