Learn More
BRAND™ seripettor™ Bottletop Dispensers
Suitable for use with bases, buffers, aqueous solutions of organic and inorganic substances, dilute non-oxidizing acids, and many polar solvents. Space-saving, low-rise design provides stability for easy one-handed operation and fit most common reagent bottles with the included adapters.
Brand: BRAND™ 4720150
139.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Includes: Two dispensing cartridges, discharge tube, filling tube, two polypropylene thread adapters (45/33 and 45/38) and operating manual
Description
Suitable for use with bases, such as ammonia and sodium hydroxide; buffers; aqueous solutions of organic and inorganic substances; dilute non-oxidizing acids; and many polar solvents.
- Fluid path consists of materials such as polyethylene, polypropylene and EPDM (ethylene-propylene-diene-rubber)
- Full scale accuracy of ≤±1.2% and reproducibility of ≤0.2%
- Fit most common reagent bottles with the included adapters
- Space-saving, low-rise design provides stability for easy one-handed operation
- With simple, clear volume adjustment for accuracy and a sturdy, supported discharge tube for safe operation
- No fragile glass parts to break
- Replaceable cylinder/piston cartridge has a wiping seal for renewability
This dispenser is not intended for use with scintillation fluid; non-polar solvents, such as xylene and most xylene substitutes; or strong, oxidizing or concentrated acids. For these liquids, choose the appropriate Dispensette™ bottletop dispenser.
Order Info
Unit disassembles in seconds for easy cleaning and maintenance. Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

Specifications
≤±1.2% | |
Up to 2.2 g/cm3 | |
Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual | |
500 mbar | |
59°F to 104°F | |
75 mm2/sec. |
Seripettor Bottletop Dispenser | |
2.5 to 25 mL | |
0.5 mL | |
≤0.2% | |
15°C to 40°C | |
1 Year |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.