Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
£232.00 - £260.00
Specifications
For Use With (Application) | Flow Cytometry |
---|---|
Product Type | Compensation Beads |
Quantity | 25 tests |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17897061
|
Invitrogen™
A54740 |
RFP BrightComp eBeads Compensation Bead Kit |
£232.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17807071
|
Invitrogen™
A54741 |
YFP BrightComp eBeads Compensation Bead Kit |
£232.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17817071
|
Invitrogen™
A54742 |
CFP BrightComp eBeads Compensation Bead Kit |
£232.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17827071
|
Invitrogen™
A54743 |
mCherry BrightComp eBeads Compensation Bead Kit |
£232.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15980213
|
Invitrogen™
A10514 |
GFP BrightComp eBeads Compensation Bead Kit |
£260.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of fluorescent protein expression• Near-identical, spectrally-matched dye to native protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
Specifications
Flow Cytometry | |
Compensation Beads | |
25 tests |