Learn More
Bromoform, 96%, stab. with ethanol, Thermo Scientific Chemicals
Description
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.
Solubility
Slightly soluble in water.
Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
Specifications
Specifications
Melting Point | 8°C to 9°C |
Density | 2.81 |
Boiling Point | 148°C to 150°C |
Flash Point | None |
Refractive Index | 1.585 |
Quantity | 250 g |
UN Number | UN2515 |
Beilstein | 1731048 |
Sensitivity | Light sensitive |
Merck Index | 14,1420 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0449442 | Certificate of Analysis | 25/01/2025 | A11904.30,A11904.0I,A11904.0B |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.