missing translation for 'onlineSavingsMsg'
Learn More
Bromophenol Blue Sodium Salt, Fisher BioReagents Product Code.: 10497573
Estimated Shipment: 09-04-2025
to see stock.
Add to basket

Bromophenol Blue Sodium Salt, Fisher BioReagents

Product Code. 10497573
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 10497573

Brand: Fisher BioReagents BP11425

Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Add to basket
Request Bulk or Custom Formats
This item is not returnable. View return policy
Bromophenol Blue Sodium Salt is commonly used as a tracking dye in electrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 300°C
CAS 34725-61-6
Clarity Pass Test
Extinction Coefficient 800g(-1) cm(-1) min.
Molecular Formula C19H9Br4NaO5S
Synonym 3,3',5,5'-Tetrabromophenolsulfonphthalein sodium salt
Identification Pass Test
InChI Key KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.94
PubChem CID 3791690
Lambda Max 585 to 595nm
Formula Weight 691.94
Loss on Drying Run and Report %
Packaging Glass Bottle
Visual Transition Interval Pass Test
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue Sodium Salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromophenol Blue Sodium Salt, Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.