missing translation for 'onlineSavingsMsg'
Learn More
Brucine dihydrate, 98%, Thermo Scientific Chemicals Product Code.: 11471980

Brucine dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 11471980
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11471980

Brand: Thermo Scientific Alfa Aesar A16251.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

Solubility
Soluble in methanol. Partially soluble Miscible with water.

Notes
Incompatible with strong oxidizing agents, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5892-11-5
MDL Number MFCD00149384
Synonym brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate

Specifications

Melting Point 176°C to 178°C
Molecular Formula C23H34N2O8
Quantity 50 g
UN Number UN1570
Solubility Information Soluble in methanol. Partially soluble Miscible with water.
InChI Key XLXHHXCMBNBDMP-BEYGIVOKSA-N
SMILES COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
Molecular Weight (g/mol) 466.53
PubChem CID 69897601
Formula Weight 430.50 (394.47 Anhydrous)
Percent Purity 98%
Chemical Name or Material Brucine dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252585 Certificate TypeCertificate of Analysis Date26/09/2024 Catalog NumberA16251.09,A16251.18
Lot Number10237927 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16251.09,A16251.18
Lot Number10197049 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16251.09,A16251.18
Lot Number10187677 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16251.09,A16251.18
Lot Number10165028 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16251.09,A16251.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brucine dihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.