Learn More
Eurofins DiscoverX BTK (E41K) Protein, Active
Active, N-terminal His6-tagged, recombinant, full-length, human BTK, for use in Kinase Assays.

£436.98 - £3335.16
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 78.4 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17360694
![]() |
Eurofins DiscoverX
14-773 |
10 μg |
£436.98
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17370694
![]() |
Eurofins DiscoverX
14-773M |
250 μg |
£3335.16
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTK (Bruton Tyrosine Kinase) is a non-receptor tyrosine kinase indispensable for B lymphocyte development, differentiation and signaling. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement. BTK also plays an important role in the function of immune cells of innate as well as adaptive immunity, as a component of the Toll-like receptors (TLR) pathway.
GenBank NM_000061
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Full-length | |
BTK (E41K), active |
78.4 kDa | |
Recombinant | |
Recombinant protein expressed in Sf21 insect cells | |
≥28% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.