missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
Brand: BTX™ 450463
This item is not returnable.
View return policy
Description
- 25-well disposable electroporation plate
- Standard multi-well formats
- Bio-compatible materials
Specifications
Transfection of Mammalian Cell, Transformation of Bacteria and Yeast, siRNA and cDNA Library Screening | |
Medical Grade polycarbonate, polished aluminum | |
Electroporation consumables | |
25-well Electroporation Plate | |
6 Pk. | |
4 mm | |
Sterile |
ECM 630HT, ECM830HT and Gemini X2 HT Electroporation Systems | |
BTX Electroporation | |
25-Well Disposable Electroporation Plate, 4 mm gap, 200 μl, pkg. of 6 | |
6 x 25-well electroporation plate, plate seals | |
0.15 in. | |
200 μL | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTX™ HT 25-well Electroporation Plates > 25-well Plate; Volume: 200 μL; Gap: 4 mm; Quantity: 6 Pk.
Spot an opportunity for improvement?Share a Content Correction