missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Bubble Sof-Tred™ Dyna-Shield™ Layer with Unique Bubble Design
Notrax Bubble Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
£43.45 - £115.00
Specifications
Color | Gray |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17752108
|
Notrax
417S0023GY |
91 cm | 60 cm |
£43.45
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
17772108
|
Notrax
417S0035GY |
150 cm | 91 cm |
£114.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
17792098
|
Notrax
417C0024GY |
1830 cm | 60 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17712108
|
Notrax
417C0036GY |
1830 cm | 91 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17732108
|
Notrax
417C0048GY |
1830 cm | 122 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Lengthwise corrugated rib pattern for sure footing
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Gray | |
Microcellular Vinyl | |
Industrial and Commercial Matting |