Learn More
Buffer Solution, pH 9.00, Reagecon™

Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 9.00 is tested at 20°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
Brand: Reagecon 10905
Description
Certified by an ISO 17025 accredited test method and fully NIST traceable. Reagecon™ Buffer Solution, pH 9.00 is tested at 20°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
0°C | |
Colorless | |
Liquid | |
pH Buffer | |
Yes |
100°C | |
9.00 ±0.01 | |
500 mL | |
Characteristic | |
Fully miscible in water |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- BUFFER SOLUTION pH 9.00 ± 0.01 a 20°C
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : boric acid, disodiumsalt
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.