Learn More
Butyl chloroformate, 98%, Thermo Scientific Chemicals
£8.40 - £125.00
Chemical Identifiers
CAS | 592-34-7 |
---|---|
MDL Number | MFCD00000650 |
InChI Key | NRDQFWXVTPZZAZ-UHFFFAOYSA-N |
Synonym | butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate |
PubChem CID | 61140 |
IUPAC Name | butyl carbonochloridate |
SMILES | CCCCOC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144150
|
Thermo Scientific Acros
180170050 |
5 mL | Glass bottle | ||||||
10532781
|
Thermo Scientific
180170250 |
25 mL | Glass bottle |
£19.50
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236140
|
Thermo Scientific Acros
180171000 |
100 mL | Glass bottle |
£24.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10512591
|
Thermo Scientific
180170010 |
1 L | Glass bottle |
£125.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
592-34-7 | |
NRDQFWXVTPZZAZ-UHFFFAOYSA-N | |
61140 | |
CCCCOC(=O)Cl |
MFCD00000650 | |
butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate | |
butyl carbonochloridate |
Specifications
592-34-7 | |
98% | |
ClCO2(CH2)3CH3 | |
MFCD00000650 | |
butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate | |
CCCCOC(=O)Cl | |
136.575 | |
136.58 | |
Liquid |
Authentic | |
C5H9ClO2 | |
1.4110 to 1.4130 (20°C, 589nm) | |
03, II, 11 | |
NRDQFWXVTPZZAZ-UHFFFAOYSA-N | |
butyl carbonochloridate | |
61140 | |
≥97.5% | |
Butyl chloroformate, 98% |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 209-750-5
missing translation for 'tsca' : TSCA
RUO – Research Use Only