Learn More
CA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16696-1-AP-150UL
Description
Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].Specifications
CA13 | |
Polyclonal | |
Unconjugated | |
CA13 | |
CA XIII, CA13, Carbonate dehydratase XIII, Carbonic anhydrase 13, carbonic anhydrase XIII, CAXIII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
377677, 499566, 71934 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N1Q1, Q9D6N1 | |
CA13, Car13 | |
CA13 Fusion Protein Ag10086 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.