Learn More
Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals
Description
Calcium bromide is a dehydrating agent used in photography, food preservatives, wood preservatives, fire retardants and freezing mixtures. It is used as an oxidizer for mercury emissions control. It is widely used for the preparation of drilling fluids, completion fluids and drilling muds formulations. It also finds application in the manufacture of mineral water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 806°C to 812°C |
Melting Point | 730°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.5% |
Solubility Information | Soluble in water,methanol,ethanol and acetone. Insoluble in dioxane,chloroform and ether. |
Formula Weight | 199.9 |
Sensitivity | Hygroscopic |
Appearance | White to off-white powder or beads |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P24L050 | Certificate of Analysis | 26/03/2025 | 087893.09,087893.18,087893.30 |
R29K039 | Certificate of Analysis | 11/06/2024 | 087893.09,087893.18,087893.30 |
R28K025 | Certificate of Analysis | 11/06/2024 | 087893.09,087893.18,087893.30 |
T03J023 | Certificate of Analysis | 10/05/2024 | 087893.09,087893.18,087893.30 |
X03J021 | Certificate of Analysis | 11/10/2023 | 087893.09,087893.18,087893.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.