missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Poly Bottle
Description
Calcium Chloride Dihydrate (White Crystals to Powder)
Calcium Chloride is suitable for use in the preparation of buffer solutions.

Specifications
Specifications
Melting Point | 175°C |
Color | Beige-White |
pH | 4.5 to 6.5 |
Physical Form | Solid |
Quantity | 250 g |
Assay Percent Range | 99.0 to 105.0 % |
Merck Index | 15, 1661 |
Formula Weight | 147.02 |
Percent Purity | 99.0 to 105.0% |
Identification | Pass Test |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Safety and Handling

Product Identifier
- Calcium Chloride Dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Calcium chloride, dihydrate, Calcium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™ >
Spot an opportunity for improvement?Share a Content Correction