missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™

£24.20 - £42.70

Chemical Identifiers

CAS 10035-04-8, 10043-52-4
Molecular Formula Ca Cl2 · 2 H2O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10306313
SDS View Documents
Fisher BioReagents
BP510-100
100 g Poly Bottle
£24.20
100g
Please to purchase this item. Need a web account? Register with us today!
10316313
SDS View Documents
Fisher BioReagents
BP510-250
250 g Poly Bottle
£40.60
250g
Please to purchase this item. Need a web account? Register with us today!
10786634
SDS View Documents Promotion Details
Fisher BioReagents
BP510-500
500 g Poly Bottle
£42.70
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium Chloride Dihydrate (White Crystals to Powder)

Calcium Chloride is suitable for use in the preparation of buffer solutions.
Specifications

Chemical Identifiers

10035-04-8, 10043-52-4
Ca Cl2 · 2 H2O

Specifications

175°C
Beige-White
Solid
Ca Cl2 · 2 H2O
147.02
Pass Test
Calcium Chloride Dihydrate
10035-04-8, 10043-52-4
4.5 to 6.5
99.0 to 105.0 %
15, 1661
99.0 to 105.0%
0.830g/cm³
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.