Learn More
Calcium hydroxide, ACS, 95.0% min, Thermo Scientific Chemicals
Description
Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materialsCalcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials

Specifications
Specifications
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 95.0% min. |
Linear Formula | Ca(OH)2 |
UN Number | UN3262 |
Merck Index | 14,1673 |
Solubility Information | Slightly soluble in water; Soluble in glycerol,acids; Insoluble in alcohol. |
IUPAC Name | calcium;dihydroxide |
Formula Weight | 74.09 |
Percent Purity | ≥95.0% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S04K033 | Certificate of Analysis | 18/06/2024 | 014662.30,014662.A1 |
S28I041 | Certificate of Analysis | 11/07/2022 | 014662.30,014662.A1 |
P26G017 | Certificate of Analysis | 12/04/2022 | 014662.30,014662.A1 |
L16Q003 | Certificate of Analysis | 08/12/2021 | 014662.30,014662.A1 |
P25H026 | Certificate of Analysis | 08/12/2021 | 014662.30,014662.A1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.