missing translation for 'onlineSavingsMsg'
Learn More
Calcium Oxide, Extra Pure, SLR, Lump, Fisher Chemical Product Code.: 10457630

Calcium Oxide, Extra Pure, SLR, Lump, Fisher Chemical

Product Code. 10457630
1kg, Plastic powder jar
Click to view available options
1kg, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10457630

Brand: Fisher Chemical C/2100/60

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 2850°C
Melting Point 2570°C
CAS 1305-78-8
Color Beige
pH 12.5
Physical Form Powder/Solid
Quantity 1 kg
Molecular Formula CaO
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES [O--].[Ca++]
IUPAC Name calcium oxidandiide
Molecular Weight (g/mol) 56.08
Formula Weight 56.08g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide lump
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Oxide, Extra Pure, SLR, Lump, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.