missing translation for 'onlineSavingsMsg'
Learn More
ε-Caprolactam, 99+%, Thermo Scientific Chemicals Product Code.: 10207313

ε-Caprolactam, 99+%, Thermo Scientific Chemicals

Product Code. 10207313
250 g, Plastic bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10207313

Brand: Thermo Scientific Acros 108232500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006936
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactamShow MoreShow Less
PubChem CID 7768
ChEBI CHEBI:28579
IUPAC Name azepan-2-one
SMILES C1CCC(=O)NCC1

Specifications

Melting Point 68°C to 71°C
Color White
Boiling Point 268°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 21, V,6, 444
Fieser 09,316
Merck Index 15, 1763
Solubility Information Solubility in water: 4560g/L (20°C). Other solubilities: freely soluble in methanol,dimethylformamide,,ethanol,ether and tetrahydrofurfuryl alcohol,soluble in chlorinated hydrocarbons,cyclohexene,petroleum fractions,chloroform,benzene
Viscosity 8.52 mPa.s (80°C)
Formula Weight 113.16
Percent Purity 99+%
Physical Form Crystals or Flakes
Chemical Name or Material ε-Caprolactam
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465704 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number108232500,108230025,108230010,108231000
Lot NumberA0464117 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number108232500,108230025,108230010,108231000
Lot NumberA0458716 Certificate TypeCertificate of Analysis Date19/12/2023 Catalog Number108232500,108230025,108230010,108231000
Lot NumberA0398423 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number108232500,108230025,108230010,108231000
Lot NumberA0447558 Certificate TypeCertificate of Analysis Date25/11/2022 Catalog Number108232500,108230025,108230010,108231000
compliance-icons
Product Identifier
  • e-Caprolactam
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ε-Caprolactam, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.