Learn More
Thermo Scientific™ CaptureSelect™ IgG4 (Hu) Affinity Matrix
Thermo Scientific™ CaptureSelect™ IgG4 (Hu) Affinity Matrix has been specifically designed for the purification and isolation of human IgG4 from complex sources such as plasma, serum, or cell culture supernatants.
£1664.00
Specifications
Binding Capacity | >15 g IgG4/L resin |
---|---|
Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
Matrix | Agarose-based, epoxide activated |
Particle Size | 65 ±10 μm |
Form | Resin Slurry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18496738
|
Thermo Scientific™
2942902005 |
5 mL |
£1664.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaptureSelect IgG4 (Hu) Affinity Matrix has been specifically designed for the purification and isolation of human IgG4 from complex sources such as plasma, serum, or cell culture supernatants. The affinity ligand selectively binds the Fc region of human IgG4, without cross-reactivity to other antibody subclasses and isotypes. The affinity resin enables a robust and efficient purification process with excellent purity obtained in a single step.
Features of this affinity matrix include:
- Highly selective for human IgG4
- No cross-reactivity with other antibody subclasses (IgA, IgD IgE, IgM) or antibody subtypes (IgG1-3)
- No cross reactivity with other species, allowing use of complex growth media containing fetal calf or horse serum
- Excellent scalability
- Non-animal-derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose-based, epoxide activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect IgG4 affinity ligand
- Ligand coupling method: epoxide coupling
- Binding capacity: >15 g IgG4/L resin depending on flow rate, column height, and contact time
- Elution conditions: 0.1 M glycine, pH 3.0
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
>15 g IgG4/L resin | |
Agarose-based, epoxide activated | |
Resin Slurry | |
Affinity Matrix |
50 to 200 cm/hr. (Up to 2 bar) | |
65 ±10 μm | |
CaptureSelect™ | |
IgG4 affinity ligand |