missing translation for 'onlineSavingsMsg'
Learn More
Carbon, activated, Norit ROW 0.8mm pellets, steam activated, Thermo Scientific Chemicals Product Code.: 11309115

Carbon, activated, Norit ROW 0.8mm pellets, steam activated, Thermo Scientific Chemicals

Product Code. 11309115
500 g
Click to view available options
Quantity:
100 g
500 g
2000 g
This item is not returnable. View return policy

Product Code. 11309115

Brand: Thermo Scientific Alfa Aesar L16334.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Carbon, activated, Norit ROW 0.8mm pellets, steam activated

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL18U017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16334.0P,L16334.22,L16334.36
Lot NumberI12T020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16334.0P,L16334.22,L16334.36
Lot NumberT15H084 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16334.0P,L16334.22,L16334.36
Lot NumberG24Y017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16334.0P,L16334.22,L16334.36
Lot NumberW30H031 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16334.0P,L16334.22,L16334.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon, activated, Norit ROW 0.8mm pellets, steam activated, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.