Learn More
Carbon, activated, Norit ROW 0.8mm pellets, steam activated, Thermo Scientific Chemicals
£26.33 - £203.01
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.011 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399105
|
Thermo Scientific Alfa Aesar
L16334.22 |
100 g |
£26.33
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309115
|
Thermo Scientific Alfa Aesar
L16334.36 |
500 g |
£63.21
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319115
|
Thermo Scientific Alfa Aesar
L16334.0P |
2000 g |
£203.01
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
Chemical Identifiers
7440-44-0 | |
12.011 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
MFCD00133992 | |
Insoluble in water. | |
[C] | |
12.011 | |
CHEBI:27594 |
C | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
Carbon, activated, Norit ROW 0.8mm pellets, steam activated |
Safety and Handling
H500
missing translation for 'einecsNumber' : 231-153-3
missing translation for 'rtecsNumber' : FF5250100
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only