Learn More
Wesbart™ Cardboard Cryoboxes
Wesbart cryogenic boxes are made from fibreboard cardboard and are water-repellent for longer durability.
Brand:  Wesbart™ A1/81
 View more versions of this product
Product Code. 17165034
  • £3.55 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Cardboard boxes are specially made for the aluminum freezer racks.
  • Dividers are available in different diameters.
Specifications
Specifications
Cardboard Box with Dividers
Cardboard
Cryo Box
Grid Divider
9 x 9
35 mm
White
Rectangular
81 Cryovials (1 to 2 mL)
Non-sterile
133 x 133 x 35 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Wesbart™ Cardboard Cryoboxes > Cryogenic Box; Dimensions: 133 x 133 x 35 mm; Holds: 81 Cryovials (1 to 2 mL); Tube Array: 9 x 9