Learn More
CD51/CD61 Monoclonal Antibody (23C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516470
Description
This antibody does not react with bovine, pig or rabbit. The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.
Specifications
CD51/CD61 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P05106, P06756, P26008 | |
ITGAV, ITGB3 | |
Osteoclasts from osteoclastomas | |
200 μg | |
Primary | |
Chicken, Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
23C6 | |
Unconjugated | |
ITGB3 | |
1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen CD61; antigen identified by monoclonal antibody L230; BDPLT16; BDPLT2; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; GT; INGRB3; integrin; integrin alpha V; integrin alpha-5 beta-3; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; Integrin beta; integrin beta 3; integrin beta 3 (Cd61); Integrin beta3; integrin beta-3; integrin subunit alpha V; integrin subunit beta 3; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGAV; Itgb3; MSK8; platelet gpIIIa; platelet membrane glycoprotein IIIa; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor alpha subunit; vitronectin receptor subunit alpha; VNRA; VTNR | |
Mouse | |
Protein A | |
RUO | |
3685, 3690, 374209, 396420 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.