Learn More
CD86 Rat anti-Mouse, FITC, Clone: GL1, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech FITC-65068-500UG
Product Code. 16841186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The CD86 antibody from Proteintech is a rat monoclonal antibody generated with lps-activated cba/ca mouse spleen b cells of mouse CD86. This antibody recognizes mouse antigen. The CD86 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
Specifications
CD86
Monoclonal
0.5 mg/mL
Rat
Primary
Mouse
Flow Cytometry
GL1
FITC
500 μg
12524
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CD86 Rat anti-Mouse, FITC, Clone: GL1, Proteintech > 500 μg