Learn More
Fisherbrand™ Traceable™ UV Light Meter
Easy-to-use meter with a wide-band UV wavelength of 320 to 390nm.
Brand: Fisherbrand™ 3253
Includes: Meter, 0.75 in. diameter (1.9cm) photo cell, 9V battery, 40 in. (1m) cable and RS-232 output for connection to IBM™-compatible computer or datalogger.
Description
Easy-to-use meter with a wide-band UV wavelength of 320 to 390nm.
Three Ranges
- 0 to 199.9μW/cm2 (0.1μW/cm2 resolution)
- 1 to 19.99mW/cm2 (0.01mW/cm22 resolution) 1 to 1.999mW/cm2 (0.001mW/cm22 resolution)
- Accuracy of ±2% full-scale plus two digits
- Convenient operation and data storage
- DATA HOLD button freezes display to capture readings
- Precise photodiode and color-correction filter provide cosine- and color-corrected measurements
- Push-button recall of highest, lowest and average readings
- RS-232 port for connectin to computer, optional Data Acquisition System 06-661-22, optional Data Logger System 06-662-72 (sold separately)
Order Info
Dimensions: 1.25L x 3W x 7 in.H (3.2 x 7.6 x 17.8cm); Weight: 9.5 oz. (269g); Optional accessories (available online): DAS-3 Data Acquisition System 11-661-22 (captures, displays on monitor and stores readings on any IBM™-compatible computer)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
UV Light Meter | |
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™. | |
3.2 cm | |
7 in. | |
0 to 199.9 uW/cm2, 1 to 19.99 mW/cm2, 1 to 1.999 mW/cm2 | |
320 to 390nm | |
7.62 cm |
±2% full-scale plus two digits | |
RS-232 port | |
Meter, 0.75 in. diameter (1.9cm) photo cell, 9V battery, 40 in. (1m) cable and RS-232 output for connection to IBM™-compatible computer or datalogger. | |
17.78 cm | |
0.1μW/cm2, 0.01mW/cm2, 1 to 1.999mW/cm2 | |
3 in. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.