Learn More
Thermo Scientific™ Restore™ PLUS Western Blot Stripping Buffer
Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
Brand: Thermo Scientific™ 46430
152.65 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific™ Restore PLUS Western Blot Stripping Buffer is an advanced formula for removing bound primary and secondary antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
Restore PLUS Buffer is an alternative formulation of the original Thermo Scientific Restore Western Blot Stripping Buffer. Restore PLUS Stripping Buffer was designed for use with antibodies that are difficult to remove from Western blots and require longer incubation times or incubation temperatures greater than 22°C with gentler formulations. High-affinity antibodies can be quickly and effectively stripped from Western blots at room temperature without removing transferred proteins, thereby allowing multiple reprobes of the target.Highlights:
Ready and easy to use – no dilution necessary; no offensive odors; store at room temperature
Compatible – use on nitrocellulose and PVDF membranes, whether still wet or already dry; works with practically any blocking buffer, enzyme conjugate and chemiluminescent substrate
Cost effective – save valuable time and samples; strip blots effectively the first time
Robust yet gentle – transferred proteins remain viable; strip the same blot up to five times
Flexible – strip and reprobe to optimize antibody concentrations or to detect a new antigen with different antibodies

Specifications
Western Blot Stripping Buffer | |
Liquid | |
Nitrocellulose, PVDF | |
500 mL |
Chemiluminescence | |
Western Blot | |
Restore™ | |
Upon receipt store at room temperature. Product shipped at ambient temperature. |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Restore(tm) PLUS Western Blot Stripping Buffer, 500 mL.
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P390-Absorb spillage to prevent material damage.
- P234-Keep only in original container.
Supplemental information
- MIXTURE LIST-Contains : Anionic Surfactant
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
87Hours
1Minutes
35Seconds
54New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.