Learn More
Invitrogen™ CellSensor™ AP-1-bla ME-180 Cell Line
CellSensor™ AP-1-bla ME-180 Cell Line
Brand: Invitrogen™ K1661
This item is not returnable.
View return policy
Description
The CellSensor™ AP-1-bla ME-180 Cell Line contains a beta-lactamase reporter gene under control of the AP-1 response element stably integrated into ME-180 cells. The cell line was created through FACS sorting of cells responsive to stimulation of the AP-1 pathway with Epidermal Growth Factor (EGF). This cell line was validated for DMSO tolerance, stimulant incubation time, substrate loading conditions, Z' and EC50 concentration of Epidermal Growth Factor. The AP-1-bla ME-180 Cell Line responds to agonist treatment as expected from the literature and can be adapted for high-throughput screening for agonists or antagonists of the AP-1 pathway with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.Specifications
Cell-based beta lactamase reporter gene | |
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
1 vial | |
Cell Line | |
CellSensor™ |
Dividing Cells | |
CellSensor™ | |
EGFR, ERK1/2, p38, MEK, MAPK pathway, c-MET, JNK, RON, RAF | |
ME-180 | |
Signaling Pathway |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
User Guide: CellSensor AP-1-bla ME-180 Cell-based Assay
Version: 2.0
File size: 162.4 kB
CellSensor®AP1-bla ME-180 Cell Line Validation Packet
Version: version 1.1
File size: 93.4 kB
Product Content Correction
Product Title
Invitrogen™ CellSensor™ AP-1-bla ME-180 Cell Line > 1 vial