Learn More
Nabertherm™ Ceramic ribbed plate
Bottom plate for protection of the furnace.
Brand: Nabertherm™ 691601097
Description
Ceramic ribbed plate
- Tmax: 1200°C
- Material: Ceramic
Specifications
Muffle Furnace LE 2 | |
1200°C | |
Ceramic | |
170 x 110 x 12.7 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.