missing translation for 'onlineSavingsMsg'
Learn More
Cerium(IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11351089

Cerium(IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11351089
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11351089

Brand: Thermo Scientific Alfa Aesar 011328.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cerium(IV) oxide is used in ceramics, and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Melting Point 2600°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 172.12
Packaging Plastic Bottle
Density 7.132 g/mL
Chemical Name or Material Cerium(IV) Oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP12L032 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog Number011328.30,011328.A1,011328.18
Lot NumberW11K015 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number011328.30,011328.A1,011328.18
Lot NumberS17K003 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number011328.30,011328.A1,011328.18
Lot NumberR10K022 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number011328.30,011328.A1,011328.18
Lot NumberQ18K028 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number011328.30,011328.A1,011328.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.