Learn More
Cesium bromide, 99% (metals basis), Thermo Scientific Chemicals
£52.76 - £225.12
Chemical Identifiers
CAS | 7787-69-1 |
---|---|
Molecular Formula | BrCs |
Molecular Weight (g/mol) | 212.81 |
MDL Number | MFCD00010954 |
InChI Key | LYQFWZFBNBDLEO-UHFFFAOYSA-M |
Synonym | cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromideShow More |
PubChem CID | 24592 |
IUPAC Name | cesium;bromide |
SMILES | [Br-].[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386858
|
Thermo Scientific Alfa Aesar
013490.18 |
50 g |
£52.76
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376858
|
Thermo Scientific Alfa Aesar
013490.30 |
250 g |
£225.12
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium bromide is used in optics as a beamsplitter component in wide-band spectrophotometers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-69-1 | |
212.81 | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
24592 | |
[Br-].[Cs+] |
BrCs | |
MFCD00010954 | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromideShow More | |
cesium;bromide |
Specifications
1300°C | |
7787-69-1 | |
Crystalline | |
BrCs | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
cesium;bromide | |
24592 | |
Odorless | |
Hygroscopic | |
Cesium bromide |
636°C | |
White | |
99% (Metals basis) | |
MFCD00010954 | |
Very soluble in water. Soluble in alcohol. Insoluble in acetone | |
[Br-].[Cs+] | |
212.81 | |
212.81 | |
1.6984 | |
4.44 g/cm3 at 20°C |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H361
missing translation for 'einecsNumber' : 232-130-0
missing translation for 'rtecsNumber' : FK9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.