Learn More
Cesium carbonate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 610°C (decomposition) |
Physical Form | Powder/Granules |
Quantity | 100 g |
Assay Percent Range | 99.9% (metals basis) |
Beilstein | 4546405 |
Solubility Information | Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide. |
IUPAC Name | dicesium;carbonate |
Formula Weight | 325.82 |
Sensitivity | Hygroscopic |
Density | 4.072 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
R20G001 | Certificate of Analysis | 02/12/2024 | 010924.14,010924.36,010924.22,010924.06 |
S22J009 | Certificate of Analysis | 28/06/2023 | 010924.14,010924.36,010924.22,010924,010924.06 |
N09I044 | Certificate of Analysis | 12/04/2022 | 010924.14,010924.36,010924.22,010924.06 |
N09I043 | Certificate of Analysis | 12/04/2022 | 010924.14,010924.36,010924.22,010924.06 |
T21H012 | Certificate of Analysis | 12/11/2021 | 010924.14,010924.36,010924.22,010924.06 |
Safety and Handling


- Caesium carbonate
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Cesium carbonate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.