Learn More
Cesium nitrate, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | (decomposition) |
Melting Point | 414°C |
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water and acetone. Slightly soluble in ethanol. |
Formula Weight | 194.91 |
Odor | Odorless |
Refractive Index | 1.55 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T29K046 | Certificate of Analysis | 06/09/2024 | 014440.22,014440.14,014440.36,014440.06 |
Y14J036 | Certificate of Analysis | 05/12/2023 | 014440.22,014440.14,014440.36,014440.06 |
S30G065 | Certificate of Analysis | 06/08/2022 | 014440.22,014440.14,014440.36,014440.06 |
E24P20 | Certificate of Analysis | 08/12/2021 | 014440.22,014440.14,014440.36,014440.06 |
I28Q010 | Certificate of Analysis | 08/12/2021 | 014440.22,014440.14,014440.36,014440.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.