missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

Accurate test kits for routine water analysis

Brand:  Lovibond™ 147040

 View more versions of this product

Product Code. 11775458


Includes:
  • Aqua Comparator Housing (D55 for L147000)
  • Aqua Comparator Disc
  • DPD No. 1 Rapid Tablets
  • DPD No. 3 Rapid Tablets
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying case
This item is not returnable. View return policy

Description

Description

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Specifications

±5%
Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
0.1 to 2mg/L
Routine water analysis
100
Chlorine Free and Total Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total > Chlorine-free, Total; 0.1-2.0mg/L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.