Learn More
Fisherbrand™ Chemical Sorbent Roll
Sorbent rolls are 2-ply, green, medium weight, bonded and double perforated chemical sorbent rolls that absorbs chemicals and hazardous liquids. One sided spunbound gives extra strength as well as less lint.
Brand: Fisherbrand™ FBEU152DPE
334.77 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Description
- Does not degrade or cause a dangerous chemical reaction with the absorbed liquid
- Green colour alerts workers the product is used for hazardous liquids
- Green colour also allows user to identify and separate hazardous waste for disposal
- Perforated down the middle and cross-wise permit the use of precisely the right amount of material - reduces waste
- Perfect for cleaning up chemical spills or for use in laboratories
Spillage, absorbent, spill, sorbent
Specifications
Saturated, Medium Viscosity: 12.5x | |
149 L | |
Chemical Sorbent | |
2 Rolls | |
46 m x 380 mm | |
380 mm |
Green | |
Polypropylene | |
Low-Lint Chemical Sorbent for Water, Oil, or Solvent Base Liquids - overspray, leaks, drips, in drum and liquid storage areas, workstations and laboratories. | |
2-ply | |
46 m |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24163Expires: 2025-04-30
Days
23Hours
6Minutes
36Seconds
41
Your input is important to us. Please complete this form to provide feedback related to the content on this product.