Learn More
Jaytec Glass™ Glass Capillary Tubes
These Glass Capillary Tubes use a liquid's properties of cohesion, adhesion and surface tension to draw liquid up the interior walls of a narrow tube.
Brand: Jaytec Glass™ CAPSPECMERC
Description
- Manufactured from industry-standard soda glass
Specifications
Soda Glass | |
Capillary Tubes | |
75 mm |
1 μL | |
0.48 to 0.5 mm | |
1000 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Jaytec Glass™ Glass Capillary Tubes > Glass Capillary Tubes; Volume: 1 μL; Length: 75 mm; Outer Diameter: 0.48 to 0.5 mm
Spot an opportunity for improvement?Share a Content Correction