Learn More
Soda Glass Capillary Tube
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
Brand:  S Murray SAMCO G119/02
 View more versions of this product
Product Code. 12807685
  • £28.47 / Pack of 100
Edge
Explore more special offers
Description
Description
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100
Specifications
Specifications
Capillary Tube
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Soda Glass Capillary Tube > Length: 100mm