Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Protects the top buffer trays from buffer spillages and salt deposits
Brand:  Cytiva 2300-10063
 View more versions of this product
Product Code. 16092032
  • £11.75 / Pack of 10
Estimated Shipment: 08-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cover the top buffer tray to protect from buffer spillages and salt deposits
  • Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
  • Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
  • Smooth, absorbent Whatman paper quickly absorbs spills
  • Laminated polyethylene layer prevents flow-through to the working surface
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Convenient and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Absorbent Liner
Paper, Laminated Polyethylene
ÄKTA Start
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Benchkote Sheet for ÄKTA Systems > ÄKTA Start, 10/Pk