Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand:  Hanna Instruments™ HI3824
 View more versions of this product
Product Code. 10480684
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
Ammonia determination in fresh water
20mL plastic beaker, color comparison cube, 20mL ammonia reagent (for fresh water), and 20mL Nessler reagent
1 Kit
Titration
Water samples
25
0 to 2.5mg/L
Ammonia Test Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Chemical Test Kit > Method: Nessler; Number of Tests: 25; Range: 0 to 2.5mg/L