missing translation for 'onlineSavingsMsg'
Learn More
Chlorobis(2-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals Product Code.: 11495640

Chlorobis(2-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals

Product Code. 11495640
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11495640

Brand: Thermo Scientific Alfa Aesar H25894.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263369-88-6
Molecular Formula C18H17ClO2P-
Molecular Weight (g/mol) 331.756
MDL Number MFCD08064037
InChI Key WVGQIZOJKMMSSK-UHFFFAOYSA-N
Synonym bis 2-methoxyphenyl chlorphosphine, acmc-20alnz, phosphinous chloride,p,p-bis 2-methoxyphenyl
PubChem CID 57371085
SMILES COC1=CC=CC=C1C2=C(P[Cl-]C=C2)C3=CC=CC=C3OC

Specifications

Melting Point 111°C to 114°C
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 280.69
Percent Purity ≥98%
Chemical Name or Material Chlorobis(2-methoxyphenyl)phosphine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10211843 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25894.06,H25894.03
Lot Number10203420 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25894.06,H25894.03
Lot Number10155660 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25894.06,H25894.03
Lot Number10155659 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25894.06,H25894.03
Lot Number10192055 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25894.06,H25894.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorobis(2-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.